Skip to Content
Merck
All Photos(1)

Documents

84368

Millipore

Salmonella Agar according to Oenöz

suitable for microbiology, NutriSelect® Plus

Synonym(s):

OENOEZ Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 15.0 g/L
bile salt mixture, 3.825 g/L
brilliant green, 0.0006 g/L
disodium hydrogen phosphate 2 hydrate, 1.0 g/L
ferric citrate, 0.5 g/L
lactose, 11.5 g/L
magnesium sulfate, 0.4 g/L
meat extract, 6.0 g/L
meat peptone, 3.8 g/L
metachrome yellow, 0.47 g/L
methyl blue, 0.25 g/L
neutral red, 0.022 g/L
L-phenylalanine, 5.0 g/L
sodium thiosulfate 5 hydrate, 4.25 g/L
starch (soluble), 0.5 g/L
sucrose, 13.0 g/L
trisodium citrate5.5 hydrate, 9.3 g/L
yeast extract, 3.0 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.0±0.2 (37 °C)

application(s)

clinical testing
environmental
food and beverages

microbiology

suitability

selective and differential for Citrobacter spp.
selective and differential for Escherichia coli
selective and differential for Klebsiella spp.
selective and differential for Proteus spp.
selective and differential for Salmonella spp.
selective and differential for Shigella spp.
selective and differential for coliforms
selective and differential for enterobacteriaceae

Application

For the cultivation of Salmonella acc. to Oenöz (1986).

Preparation Note

Dissolve 80.3 g in 1 litre water, boil to suspend. Do NOT autoclave.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Onöz et al.
Zentralblatt fur Bakteriologie, Parasitenkunde, Infektionskrankheiten und Hygiene. Erste Abteilung Originale. Reihe A: Medizinische Mikrobiologie und Parasitologie, 240(1), 16-21 (1978-01-01)
A new medium for the cultivation of Salmonella is presented which enables an immediate distinction of Proteus bacteria and Citrobacter. The time needed for diagnosis is reduced, expenditure on staff and material will be reduced. The "positive" salmonella yield from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service