Skip to Content
Merck
All Photos(1)

Documents

82612

Sigma-Aldrich

Pyrazinecarboxamide

≥99.0% (N)

Synonym(s):

Pyrazinamide, Pyrazinoic acid amide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N3O
CAS Number:
Molecular Weight:
123.11
Beilstein:
112306
EC Number:
MDL number:
UNSPSC Code:
51101500
PubChem Substance ID:

Assay

≥99.0% (N)

mp

189-191 °C (lit.)
189-191 °C

antibiotic activity spectrum

mycobacteria

SMILES string

NC(=O)c1cnccn1

InChI

1S/C5H5N3O/c6-5(9)4-3-7-1-2-8-4/h1-3H,(H2,6,9)

InChI key

IPEHBUMCGVEMRF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service