Skip to Content
Merck
All Photos(1)

Key Documents

80910

Sigma-Aldrich

Pivaloyl chloride

purum, ≥98.0% (GC)

Synonym(s):

Trimethylacetyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCOCl
CAS Number:
Molecular Weight:
120.58
Beilstein:
385668
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

vapor density

>1 (vs air)

vapor pressure

36 mmHg ( 20 °C)

grade

purum

Assay

≥98.0% (GC)

form

liquid

refractive index

n20/D 1.412 (lit.)
n20/D 1.412

bp

105-106 °C (lit.)

density

0.980 g/mL at 20 °C
0.979 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)C(Cl)=O

InChI

1S/C5H9ClO/c1-5(2,3)4(6)7/h1-3H3

InChI key

JVSFQJZRHXAUGT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

K Noda et al.
International journal of peptide and protein research, 36(2), 197-200 (1990-08-01)
A series of p-nitroanilides of t-butyloxycarbonylamino acids, including Boc-Trp, Boc-Asn, Boc-Gln, Boc-Ser(But), Boc-Thr(But), Boc-Asp(OBut), Boc-Lys(TFA), and Boc-His(Boc), were prepared conveniently by the mixed anhydride method using 2,2-dimethylpropanoic chloride (pivaloyl chloride). The products were obtained in 40-60% yields after purification by
Srdanka Tomić et al.
Carbohydrate research, 338(6), 491-494 (2003-04-02)
A series of methyl O-pivaloyl-alpha-D-mannopyranosides was synthesized using pivaloyl chloride in pyridine. The 3,6-di-O-pivaloyl derivative 6 undergoes intramolecular transesterification in neutral conditions (buffer, pH 7.2) to give its 2,6-di-O-pivaloyl analogue 5. The course of this migration was followed using 14C-labelled
Susannah Sigurdsson et al.
Nucleosides, nucleotides & nucleic acids, 22(1), 1-12 (2003-04-24)
A kinetic study on the reactions of pivaloyl chloride with a nucleoside H-phosphonate (bisacylphosphite formation), or a protected nucleoside with a free 5'-OH (5'-O-pivaloylation) has been carried out in the presence of differently substituted pyridines. The bisacylphosphite formation is a
Michal Sobkowski et al.
Nucleosides, nucleotides & nucleic acids, 29(8), 628-645 (2010-07-28)
Efficiency and stereoselectivity of condensations of ribonucleoside 3'-H-phosphonates with ethanol promoted by pivaloyl chloride were investigated as a function of tertiary amines used. Side reactions leading to an increased demand for the condensing agent were identified as derived from an
4'-pivaloyl substituted thymidine as a precursor for the thymyl radical: an EPR spectroscopic study.
Olav Schiemann et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 5(2), 270-274 (2004-03-25)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service