Skip to Content
Merck
All Photos(1)

Key Documents

74010

Sigma-Aldrich

Nitrosyl tetrafluoroborate

purum, ≥98.0% (T)

Synonym(s):

Nitrosonium tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NOBF4
CAS Number:
Molecular Weight:
116.81
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

grade

purum

Assay

≥98.0% (T)

reaction suitability

reagent type: oxidant

storage temp.

2-8°C

SMILES string

N#[O+].F[B-](F)(F)F

InChI

1S/BF4.NO/c2-1(3,4)5;1-2/q-1;+1

InChI key

KGCNVGDHOSFKFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service