Skip to Content
Merck
All Photos(1)

Key Documents

73995

Sigma-Aldrich

Nitrosulfonazo III

p.a., indicator

Synonym(s):

2,7-Bis(4-nitro-2-sulfophenylazo)chromotropic acid tetrasodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H10N6Na4O18S4
CAS Number:
Molecular Weight:
866.56
Beilstein:
1899945
EC Number:
MDL number:
UNSPSC Code:
12171500

grade

indicator grade
indicator
p.a.

mp

222 °C (lit.)

SMILES string

[Na+].[Na+].Oc1c(\N=N\c2ccc(cc2S(O)(=O)=O)[N+]([O-])=O)c(cc3cc(c(\N=N\c4ccc(cc4S(O)(=O)=O)[N+]([O-])=O)c(O)c13)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C22H14N6O18S4.2Na/c29-21-18-9(5-16(49(41,42)43)19(21)25-23-12-3-1-10(27(31)32)7-14(12)47(35,36)37)6-17(50(44,45)46)20(22(18)30)26-24-13-4-2-11(28(33)34)8-15(13)48(38,39)40;;/h1-8,29-30H,(H,35,36,37)(H,38,39,40)(H,41,42,43)(H,44,45,46);;/q;2*+1/p-2/b25-23+,26-24+;;

InChI key

CKLNXXLIRDZLQS-SPBSJSFYSA-L

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service