Skip to Content
Merck
All Photos(1)

Documents

73538

Supelco

DL-4-Hydroxy-3-methoxymandelic acid

analytical standard

Synonym(s):

α,4-Dihydroxy-3-methoxybenzeneacetic acid, VMA, Vanillomandelic acid, Vanillylmandelic acid, Vanilmandelic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(OCH3)CH(OH)CO2H
CAS Number:
Molecular Weight:
198.17
Beilstein:
2213227
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)
98.0-102.0% (T)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

132-134 °C (dec.) (lit.)

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

OC1=C(OC)C=C(C(O)C(O)=O)C=C1

InChI

1S/C9H10O5/c1-14-7-4-5(2-3-6(7)10)8(11)9(12)13/h2-4,8,10-11H,1H3,(H,12,13)

InChI key

CGQCWMIAEPEHNQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roger L Novack et al.
Ophthalmology, 122(4), 796-802 (2014-12-02)
The evaluation of the safety and preliminary efficacy of 125 μg ocriplasmin intravitreal injection in patients with focal vitreomacular adhesion (VMA) and exudative age-related macular degeneration (AMD). Randomized, sham-injection controlled, double-masked, multicenter, phase II trial. A total of 100 patients
Simona Codruţa Cobzac et al.
Journal of chromatographic science, 52(9), 1095-1103 (2013-10-29)
The retention behavior for a series of amine neurotransmitters, their precursors, metabolites and structurally related drugs has been investigated in reversed-phase thin-layer chromatography using RP-18, RP-8, RP-2, CN and Diol stationary phases and mixtures of phosphate buffer and methanol as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service