Skip to Content
Merck
All Photos(1)

Key Documents

68620

Sigma-Aldrich

(R)-(+)-N,α-Dimethylbenzylamine

purum, ≥96.0% (sum of enantiomers, GC)

Synonym(s):

(R)-(+)-N-Methyl-1-phenylethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH3)NHCH3
CAS Number:
Molecular Weight:
135.21
Beilstein:
4350160
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

grade

purum

Assay

≥96.0% (sum of enantiomers, GC)

optical activity

[α]20/D +74±4°, c = 2% in chloroform

refractive index

n20/D 1.511 (lit.)
n20/D 1.511

bp

74-76 °C/11 mmHg (lit.)

density

0.924 g/mL at 20 °C (lit.)

SMILES string

CN[C@H](C)c1ccccc1

InChI

1S/C9H13N/c1-8(10-2)9-6-4-3-5-7-9/h3-8,10H,1-2H3/t8-/m1/s1

InChI key

RCSSHZGQHHEHPZ-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Chiral secondary amine used as auxiliary, e.g. detn. of absolute configuration of carboxylic acids; Asymmetric addition of cuprates to enones; Preparation of chiral thiophosphonate

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

141.8 °F - closed cup

Flash Point(C)

61 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M.S. McQueney et al.
The Journal of Organic Chemistry, 56, 7121-7121 (1991)
B.E. Rossiter et al.
Tetrahedron Letters, 31, 965-965 (1990)
O. Cervinka et al.
Collection of Czechoslovak Chemical Communications, 51, 404-404 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service