Skip to Content
Merck
All Photos(1)

Key Documents

676888

Sigma-Aldrich

Chloroform

ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer

Synonym(s):

Methylidyne trichloride, Trichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHCl3
CAS Number:
Molecular Weight:
119.38
Beilstein:
1731042
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:

grade

ACS reagent

vapor density

4.1 (vs air)

vapor pressure

160 mmHg ( 20 °C)

Assay

≥99.8%

contains

0.5-1.0% ethanol as stabilizer

refractive index

n20/D 1.445 (lit.)

bp

60.5-61.5 °C (lit.)

mp

−63 °C (lit.)

density

1.48 g/mL at 25 °C
1.492 g/mL at 25 °C (lit.)

SMILES string

ClC(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H

InChI key

HEDRZPFGACZZDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiu-Hao Chen et al.
Biophysical journal, 97(10), 2794-2802 (2009-11-18)
X-ray reflectivity measurements are used to determine the configuration of the C2 domain of protein kinase Calpha (PKCalpha-C2) bound to a lipid monolayer of a 7:3 mixture of 1-stearoyl-2-oleoyl-sn-glycero-3-phosphocholine and 1-stearoyl-2-oleoyl-sn-glycero-3-phosphoserine supported on a buffered aqueous solution. The reflectivity is
Carsten Schwarz et al.
Nucleic acids research, 37(10), 3215-3229 (2009-03-27)
Despite being plagued by heavily degraded DNA in palaeontological remains, most studies addressing the state of DNA degradation have been limited to types of damage which do not pose a hindrance to Taq polymerase during PCR. Application of serial qPCR
C Nagelreiter et al.
International journal of pharmaceutics, 491(1-2), 162-169 (2015-06-29)
The tape stripping method is a very important tool for dermopharmacokinetic experiments in vitro and the accurate measurement of the removed corneocytes is key for a reliable calculation of a drug's skin penetration behavior. Therefore, various methods to quantify the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service