61796
Lead Ionophore VIII
Selectophore™, function tested, ≥95.0% (HPLC)
Synonym(s):
2,7-Bis(1,1-dimethylethyl)-9,9-dimethyl-N,N′-bis[4-(10,15,20-triphenyl-21H,23H-porphin-5-yl)phenyl]-9H-xanthene-4,5-dicarboxamide
About This Item
product line
Selectophore™
Assay
≥95.0% (HPLC)
quality
function tested
SMILES string
CC(C)(C)c1cc(C(=O)Nc2ccc(cc2)-c3c4ccc(n4)c(-c5ccccc5)c6ccc([nH]6)c(-c7ccccc7)c8ccc(n8)c(-c9ccccc9)c%10ccc3[nH]%10)c%11Oc%12c(cc(cc%12C(C)(C)c%11c1)C(C)(C)C)C(=O)Nc%13ccc(cc%13)-c%14c%15ccc(n%15)c(-c%16ccccc%16)c%17ccc([nH]%17)c(-c%18ccccc%18)c%19ccc(n%19)c(-c%20ccccc%20)c%21ccc%14[nH]%21
InChI
1S/C113H88N10O3/c1-111(2,3)75-63-79(109(124)114-77-43-39-73(40-44-77)105-95-59-55-91(120-95)101(69-31-19-11-20-32-69)87-51-47-83(116-87)99(67-27-15-9-16-28-67)84-48-52-88(117-84)102(70-33-21-12-22-34-70)92-56-60-96(105)121-92)107-81(65-75)113(7,8)82-66-76(112(4,5)6)64-80(108(82)126-107)110(125)115-78-45-41-74(42-46-78)106-97-61-57-93(122-97)103(71-35-23-13-24-36-71)89-53-49-85(118-89)100(68-29-17-10-18-30-68)86-50-54-90(119-86)104(72-37-25-14-26-38-72)94-58-62-98(106)123-94/h9-66,116,118,121,123H,1-8H3,(H,114,124)(H,115,125)/b99-83-,99-84-,100-85-,100-86-,101-87-,101-91-,102-88-,102-92-,103-89-,103-93-,104-90-,104-94-,105-95-,105-96-,106-97-,106-98-
InChI key
JIKRGXBBUHYWQH-YZVDOJMOSA-N
General description
Packaging
Legal Information
related product
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service