Skip to Content
Merck
All Photos(1)

Key Documents

61130

Millipore

Kundrat Agar

suitable for microbiology

Synonym(s):

Test Agar for the Residue Test according to Kundrat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

sterility

non-sterile

Quality Level

form

solid

shelf life

limited shelf life, expiry date on the label

composition

agar, 10 g/L
bromocresol purple, 0.016 g/L
casein peptone, 7.8 g/L
gelatin, 4 g/L
D(+)-glucose, 1 g/L
meat peptone, 7.8 g/L
sodium chloride, 3 g/L
starch, 4 g/L
yeast extract, 2.8 g/L

final pH

6.8±0.2 (25 °C)

application(s)

food and beverages
microbiology

suitability

nonselective and differential for Bacillus spp.

Application

For the routine qualitative detection of residues from antibiotics, sulfonamides and other chemotherapeutical agents in animal-derived food according to Kundrat.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W. Kundrat
Z. Anal. Chem., 243, 624-624 (1968)
ibid.
Fleischwirtsch., 52, 485-485 (1972)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service