59573
Acid Red 33
analytical standard
Synonym(s):
5-Amino-4-hydroxy-3-phenylazo-2,7-naphthalenedisulfonic acid disodium salt, Acid Red 33 disodium salt, Disodium 5-amino-4-hydroxy-3-(phenylazo)-naphthalene-2,7-disulfonate, Food Red 12
Select a Size
Select a Size
About This Item
Recommended Products
grade
analytical standard
Quality Level
Assay
≥97.0% (HPLC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
cleaning products
cosmetics
food and beverages
personal care
format
neat
SMILES string
O=S(C1=CC2=CC(S(=O)([O-])=O)=CC(N)=C2C(O)=C1/N=N/C3=CC=CC=C3)([O-])=O.[Na+].[Na+]
InChI
1S/C16H13N3O7S2.2Na/c17-12-8-11(27(21,22)23)6-9-7-13(28(24,25)26)15(16(20)14(9)12)19-18-10-4-2-1-3-5-10;;/h1-8,20H,17H2,(H,21,22,23)(H,24,25,26);;/q;2*+1/p-2/b19-18+;;
InChI key
LQJVOKWHGUAUHK-BUFQOAPZSA-L
Application
Recommended products
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service