Skip to Content
Merck
All Photos(1)

Key Documents

57225

Sigma-Aldrich

Indole-3-carboxylic acid

purum, ≥98.0% (T)

Synonym(s):

β-Indolylcarboxylic acid, 3-Carboxyindole, 3-Indole formic acid, 3-Indolylcarboxylic acid, Indole-β-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7NO2
CAS Number:
Molecular Weight:
161.16
Beilstein:
129435
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥98.0% (T)

mp

232-234 °C (dec.) (lit.)

SMILES string

OC(=O)c1c[nH]c2ccccc12

InChI

1S/C9H7NO2/c11-9(12)7-5-10-8-4-2-1-3-6(7)8/h1-5,10H,(H,11,12)

InChI key

KMAKOBLIOCQGJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for preparation of:
  • Anticancer agents
  • Derivatives of amino acids and peptides
  • Serotonin 5-HT4 receptor antagonists
  • Primary acylureas
  • Inhibitors of Gli1-mediated transcription in the Hedgehog pathway
  • Serotonin 5-HT6 antagonists
  • Very Late Antigen-4 (VLA-4) sntagonists
  • EphB3 receptor tyrosine kinase inhibitors
  • Potential Therapeutic Agent for Alzheimer′s Disease
  • Vinyl ester pseudotripeptide proteasome inhibitors

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mu-Yang Wang et al.
Journal of integrative plant biology, 54(7), 471-485 (2012-05-26)
Camalexin (3-thiazol-2'-yl-indole) is the major phytoalexin found in Arabidopsis thaliana. Several key intermediates and corresponding enzymes have been identified in camalexin biosynthesis through mutant screening and biochemical experiments. Camalexin is formed when indole-3-acetonitrile (IAN) is catalyzed by the cytochrome P450
M U Ahmad et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 23(9), 841-847 (1985-09-01)
The nitrosation of gramine, a tertiary amine alkaloid present in barley malt, was carried out by reaction with sodium nitrite in buffered acetic acid (pH 3.4) for 1 hr at room temperature. Two major non-volatile products of the nitrosation reaction
J Hagemeier et al.
Proceedings of the National Academy of Sciences of the United States of America, 98(2), 753-758 (2001-01-03)
The chemical structures and accumulation kinetics of several major soluble as well as wall-bound, alkali-hydrolyzable compounds induced upon infection of Arabidopsis thaliana leaves with Pseudomonas syringae pathovar tomato were established. All identified accumulating products were structurally related to tryptophan. Most
Jin-Mo Ku et al.
The Journal of organic chemistry, 72(21), 8115-8118 (2007-09-20)
An enantioselective synthetic method for (-)-cis-clavicipitic acid (1) was reported. 1 was obtained in 10 steps (99% ee and 20% overall yield) from 1H-indole-3-carboxylic acid methyl ester (9) via asymmetric phase-transfer catalytic alkylation and diastereoselective Pd(II)-catalyzed intramolecular aminocyclization as key
Toyokazu Yoshida et al.
Bioscience, biotechnology, and biochemistry, 66(11), 2388-2394 (2003-01-01)
After enrichment culture with indole-3-carboxylate in static culture, a novel reversible decarboxylase, indole-3-carboxylate decarboxylase, was found in Arthrobacter nicotianae FI1612 and several molds. The enzyme reaction was examined in resting-cell reactions with A. nicotianae FI1612. The enzyme activity was induced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service