Skip to Content
Merck
All Photos(1)

Documents

55493

Sigma-Aldrich

Hydroxylamine-O-sulfonic acid

purum, ≥95.0% (RT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NOSO3H
CAS Number:
Molecular Weight:
113.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥95.0% (RT)

mp

210 °C (dec.) (lit.)
~225 °C (dec.)

storage temp.

2-8°C

SMILES string

NOS(O)(=O)=O

InChI

1S/H3NO4S/c1-5-6(2,3)4/h1H2,(H,2,3,4)

InChI key

DQPBABKTKYNPMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

may contain isopropyl ether

Other Notes

Review of its use in organic chemistry

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R.G. Wallace
Organic preparations and procedures international, 14, 269-269 (1982)
K Sakano et al.
Mutation research, 479(1-2), 101-111 (2001-07-27)
2-Nitropropane (2-NP), a widely used industrial solvent, is carcinogenic to rats. To clarify the mechanism of carcinogenesis by 2-NP, we investigated DNA damage by 2-NP metabolites, N-isopropylhydroxylamine (IPHA) and hydroxylamine-O-sulfonic acid (HAS), using 32P-5'-end-labelled DNA fragments obtained from genes that
K Kohda et al.
Nucleic acids symposium series, (17)(17), 145-148 (1986-01-01)
Amination of guanosine (Guo) with 2,4-dinitrophenoxyamine in aqueous DMF gave 7-amino-Guo, which was readily converted to 8,5'-O-cyclo-Guo, and 8-hydroxy-Guo. Deoxyguanosine (dG) gave only deglycosylated 7-amino-G under the same reaction condition. Aminations of Guo and dG with hydroxylamine-O-sulfonic acid above pH
Aleesha M McCormick et al.
Acta biomaterialia, 28, 33-44 (2015-09-24)
Immobilization of axon guidance cues offers a powerful tissue regenerative strategy to control the presentation and spatial location of these biomolecules. We use our previously developed immobilization strategy to specifically tether recombinant biotinylated nerve growth factor (bNGF) and biotinylated semaphorin3A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service