Skip to Content
Merck
All Photos(1)

Key Documents

50655

Sigma-Aldrich

Glyoxal trimer dihydrate

Synonym(s):

Ethanedial trimer dihydrate, Hexahydro[1,4]dioxino[2,3-b]-1,4-dioxin-2,3,6,7-tetrol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HC(O)C(O)H)3 · 2H2O
CAS Number:
Molecular Weight:
210.14
Beilstein:
3542160
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

SMILES string

[H]O[H].[H]O[H].OC1OC2OC(O)C(O)OC2OC1O

InChI

1S/C6H10O8/c7-1-2(8)12-6-5(11-1)13-3(9)4(10)14-6/h1-10H

InChI key

BDSPTFQIOAEIII-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in the synthesis of:
  • Aza-analogs of 1,4-naphthoquinones as effectors of plasmodial thioredoxin and glutathione reductase
  • Substituted side-bridged cyclam derivatives
  • Furandicarboxylic acid di-methyl ester derivatives

Other Notes

This form of glyoxal is composed of 3 moles of glyoxal and 2 moles of water in a relatively stable configuration.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>239.0 °F

Flash Point(C)

> 115 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service