Skip to Content
Merck
All Photos(1)

Key Documents

48205

Sigma-Aldrich

Fusaric acid

≥99.0% (TLC), from Gibberella fujikuroi

Synonym(s):

5-Butylpicolinic acid, 5-Butylpyridine-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13NO2
CAS Number:
Molecular Weight:
179.22
Beilstein:
125804
EC Number:
MDL number:
UNSPSC Code:
12352200

biological source

Gibberella fujikuroi

Assay

≥99.0% (TLC)

mp

96-100 °C

solubility

ethanol: 50 mg/mL, clear, faintly yellow

Mode of action

enzyme | inhibits

storage temp.

−20°C

SMILES string

CCCCc1ccc(nc1)C(O)=O

InChI

1S/C10H13NO2/c1-2-3-4-8-5-6-9(10(12)13)11-7-8/h5-7H,2-4H2,1H3,(H,12,13)

InChI key

DGMPVYSXXIOGJY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: pyridine

Other Notes

Inhibits dopamine β-hydroxylase activity

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fusaric (5-butylpicolinic) acid, an inhibitor of dopamine beta-hydroxylase, affects serotonin and noradrenaline.
H Hidaka
Nature, 231(5297), 54-55 (1971-05-07)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service