Skip to Content
Merck
All Photos(1)

Key Documents

46476

Supelco

Nandrolone

VETRANAL®, analytical standard

Synonym(s):

19-Nortestosterone, 17β-Hydroxy-19-norandrost-4-en-3-one, 17β-Hydroxy-4-estren-3-one, 19-Norandrostenolone, 4-Estren-17β-ol-3-one, Nadrolone, Nandrolone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26O2
CAS Number:
Molecular Weight:
274.40
Beilstein:
2055849
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

drug control

USDEA Schedule IIIN; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1CC[C@@H]2O

InChI

1S/C18H26O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h10,13-17,20H,2-9H2,1H3/t13-,14+,15+,16-,17-,18-/m0/s1

InChI key

NPAGDVCDWIYMMC-IZPLOLCNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Segura et al.
Forensic science international, 107(1-3), 347-359 (2000-02-26)
Detection of anabolic steroids in hair samples has been possible only in fatal cases or in cases of high-continuous dosages. In order to verify the possibility of detecting an acute administration, a sensitive and specific assay has been developed for
Terence F McGrath et al.
Analytical and bioanalytical chemistry, 405(4), 1171-1179 (2012-11-16)
Unregulated growth promoter use in food-producing animals is an issue of concern both from food safety and animal welfare perspectives. However, the monitoring of such practices is analytically challenging due to the concerted actions of users to evade detection. Techniques
Pieter Van Renterghem et al.
Analytica chimica acta, 768, 41-48 (2013-03-12)
Due to their performance enhancing properties, use of anabolic steroids (e.g. testosterone, nandrolone, etc.) is banned in elite sports. Therefore, doping control laboratories accredited by the World Anti-Doping Agency (WADA) screen among others for these prohibited substances in urine. It
P Kintz et al.
Journal of pharmaceutical and biomedical analysis, 24(5-6), 1125-1130 (2001-03-15)
A sensitive, specific and reproducible method for the quantitative determination of nandrolone in human hair has been developed. The sample preparation involved a decontamination step of the hair with methylene chloride. The hair sample (about 100 mg) was solubilized in
Katrien Venken et al.
Bone, 36(4), 663-670 (2005-03-24)
This study reports the preclinical evaluation of the bone and muscle protective potential of the synthetic androgen 7alpha-methyl-19-nortestosterone (MENTtrade mark), as assessed in the aged orchidectomized rat model. Aged (13-month-old) orchidectomized Wistar rats were treated with different doses of MENT

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service