Skip to Content
Merck
All Photos(1)

Key Documents

44891

Supelco

2,3,4,6-Tetra-O-pivaloyl-β-D-glucopyranosyl isothiocyanate

for chiral derivatization, LiChropur, ≥95.0% (HPLC)

Synonym(s):

PGITC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H43NO9S
CAS Number:
Molecular Weight:
557.70
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

derivatization grade ((chiral))
for chiral derivatization

Quality Level

product line

ChiraSelect

Assay

≥95.0% (HPLC)

optical purity

enantiomeric ratio: ≥99.5:0.5

quality

LiChropur

SMILES string

CC(C)(C)C(=O)OC[C@H]1O[C@@H](N=C=S)[C@H](OC(=O)C(C)(C)C)[C@@H](OC(=O)C(C)(C)C)[C@@H]1OC(=O)C(C)(C)C

InChI

1S/C27H43NO9S/c1-24(2,3)20(29)33-13-15-16(35-21(30)25(4,5)6)17(36-22(31)26(7,8)9)18(19(34-15)28-14-38)37-23(32)27(10,11)12/h15-19H,13H2,1-12H3/t15-,16-,17+,18-,19-/m1/s1

InChI key

QCEYERYMOHEQHK-UJWQCDCRSA-N

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

ChiraSelect is a trademark of Sigma-Aldrich Co. LLC
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 2, 3, 4, 6-tetra-O-pivaloyl-beta-D-glucopyranosyl Isothiocyanate.
Zhou, G., X. Liu, and C. Yang.
JOURNAL-SHANDONG UNIVERSITY OF SCIENCE AND TECHNOLOGY NATURAL SCIENCE, 25.4, 60-60 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service