Skip to Content
Merck
All Photos(1)

Key Documents

429627

Sigma-Aldrich

Formaldehyde-2,4-dinitrophenylhydrazone

analytical standard, for environmental analysis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=NNHC6H3(NO2)2
CAS Number:
Molecular Weight:
210.15
Beilstein:
750330
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

grade

analytical standard, for environmental analysis

Assay

99%

form

solid

mp

153-156 °C (lit.)

application(s)

environmental
hematology
histology

storage temp.

room temp

SMILES string

[O-][N+](=O)c1ccc(NN=C)c(c1)[N+]([O-])=O

InChI

1S/C7H6N4O4/c1-8-9-6-3-2-5(10(12)13)4-7(6)11(14)15/h2-4,9H,1H2

InChI key

UEQLSLWCHGLSML-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Teresa M Lamb et al.
PloS one, 6(11), e27149-e27149 (2011-11-17)
MAPK signal transduction pathways are important regulators of stress responses, cellular growth, and differentiation. In Neurospora, the circadian clock controls rhythms in phosphorylation of the p38-like MAPK (OS-2); however, the mechanism for this regulation is not known. We show that
Determination of formaldehyde in hair creams by gas chromatography-mass spectrometry.
Fabiana A Lobo et al.
Drug testing and analysis, 7(9), 848-852 (2015-05-02)
Ikuo Ueta et al.
Analytical and bioanalytical chemistry, 407(3), 899-905 (2014-09-10)
A novel miniaturized sample extraction capillary was developed to provide a simple and sensitive method for analyzing gaseous formaldehyde (FA) using conventional high-performance liquid chromatography (HPLC). The extraction capillary was prepared by packing silica gel particles in a stainless steel

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service