Skip to Content
Merck
All Photos(1)

Documents

402893-M

Sigma-Aldrich

1-Propanol

ACS reagent, ≥99.5%

Synonym(s):

Propyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2OH
CAS Number:
Molecular Weight:
60.10
Beilstein:
1098242
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

vapor density

2.1 (vs air)

vapor pressure

10 mmHg ( 147 °C)
14.9 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

700 °F

expl. lim.

13.7 %

impurities

≤0.0004 meq/g Titr. acid
≤0.01% ethanol
≤0.01% methanol
≤0.03% Carbonyl compounds (as C2H5CHO)
≤0.05% CH3CHOHCH3
≤0.05% isopropanol
≤0.2% water

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.384 (lit.)

pH

8.5 (20 °C, 200 g/L)

bp

97 °C (lit.)

mp

−127 °C (lit.)

solubility

H2O: passes test
water: soluble

density

0.804 g/mL at 25 °C (lit.)

SMILES string

CCCO

InChI

1S/C3H8O/c1-2-3-4/h4H,2-3H2,1H3

InChI key

BDERNNFJNOPAEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Propanol may be used as a solvent in the following processes:
  • Synthesis of ZnO (zinc oxide) thin films.
  • Growth of rubrene (5,6,11,12-tetraphenyltetracene) single crystals.
  • Synthesis of polyaniline microspheres and nanotubes.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Deng et al.
Metabolic engineering, 13(5), 570-577 (2011-07-16)
Biofuel production from renewable resources can potentially address lots of social, economic and environmental issues but an efficient production method has yet to be established. Combinations of different starting materials, organisms and target fuels have been explored with the conversion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service