Skip to Content
Merck
All Photos(1)

Key Documents

38753

Supelco

Bisphenol AP

analytical standard

Synonym(s):

4,4′-(1-Phenylethylidene)bisphenol, Bisphenol AP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C(C6H5)(C6H4OH)2
CAS Number:
Molecular Weight:
290.36
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

188-191 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CC(c1ccccc1)(c2ccc(O)cc2)c3ccc(O)cc3

InChI

1S/C20H18O2/c1-20(15-5-3-2-4-6-15,16-7-11-18(21)12-8-16)17-9-13-19(22)14-10-17/h2-14,21-22H,1H3

InChI key

VOWWYDCFAISREI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bisphenol AP (BPAP), an analog of bisphenol A (BPA), is an endocrine disruptor among the bisphenol analogs and one of the most used products in contact materials. It is reported to exert genotoxic potentials, greater than BPA.

Application

Bisphenol AP may be used as an analytical reference standard for the quantification of the analyte in:
  • Urine samples and beverages using liquid chromatography-tandem mass spectrometry (LC-MS/MS).
  • Wastewaters using gas-chromatography coupled to mass spectrometry (GC-MS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of bisphenols in beverages by mixed-mode solid-phase extraction and liquid chromatography coupled to tandem mass spectrometry
Regueiro J and Wenzl T
Journal of Chromatography A, 1422, 230-238 (2015)
The occurrence and source identification of bisphenol compounds in wastewaters
Cesen M, et al.
The Science of the Total Environment, 616, 744-752 (2018)
Genotoxic potentials and related mechanisms of bisphenol A and other bisphenol compounds: a comparison study employing chicken DT40 cells
Lee S, et al.
Chemosphere, 93(2), 434-440 (2013)
Determination of bisphenol A and related substitutes/analogues in human breast milk using gas chromatography-tandem mass spectrometry
Deceuninck Y, et al.
Analytical and Bioanalytical Chemistry, 407(9), 2485-2497 (2015)
Bruno Alves Rocha et al.
Talanta, 154, 511-519 (2016-05-08)
In this study, a novel method combining dispersive liquid-liquid microextraction (DLLME) and fast liquid chromatography coupled to mass spectrometry (LC-MS/MS) was developed and validated for the extraction and determination of bisphenol A (BPA) and six bisphenol analogues, namely bisphenol S

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service