Skip to Content
Merck
All Photos(1)

Key Documents

37426

Supelco

AQUANAL-plus magnesium (Mg) 100-1500 mg/L

test set with color comparator and check solution

Sign Into View Organizational & Contract Pricing


About This Item

Beilstein:
4948473
MDL number:
UNSPSC Code:
41113319
PubChem Substance ID:

usage

sufficient for 60 tests

measuring range

10-1000 mg/L (photometric, Spectro 3)
100-1500 mg/L (color chart)

application(s)

environmental
general analytical

SMILES string

[Mg]

InChI

1S/Mg

InChI key

FYYHWMGAXLPEAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aquanal kits can be used for the determination of hardness and phosphate ammonia, nitrate, and nitrite concentrations.

Analysis Note

Method
Formation of a red dye with reagent according to Mann and Yoe

Gradation (color chart)
0 - 100 - 200 - 300 - 500 - 1000 - 1500 mg/L (Mg2+)

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Removal of Mercury from Chloralkali Electrolysis Wastewater by a Mercury-Resistant Pseudomonas putidaStrain.
Canstein V
Applied and Environmental Microbiology, 65 (12), 5279-5284 (1999)
Zhichao Liu et al.
PLoS computational biology, 7(12), e1002310-e1002310 (2011-12-24)
Drug-induced liver injury (DILI) is a significant concern in drug development due to the poor concordance between preclinical and clinical findings of liver toxicity. We hypothesized that the DILI types (hepatotoxic side effects) seen in the clinic can be translated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service