Skip to Content
Merck
All Photos(1)

Key Documents

36747

Supelco

3-Chlorophenol

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4OH
CAS Number:
Molecular Weight:
128.56
Beilstein:
1634401
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.563 (lit.)

bp

214 °C (lit.)

mp

31-34 °C (lit.)

density

1.218 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Oc1cccc(Cl)c1

InChI

1S/C6H5ClO/c7-5-2-1-3-6(8)4-5/h1-4,8H

InChI key

HORNXRXVQWOLPJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

248.0 °F - closed cup

Flash Point(C)

120 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S H Lin et al.
Waste management (New York, N.Y.), 22(6), 595-603 (2002-09-07)
Experimental investigations were conducted on the adsorption characteristics of phenol and m-chlorophenol by organobentonites. The organobentonites were prepared by modifying natural bentonite with various quaternary ammonium salts including tetramethylammonium bromide, hexadecyltrimethylammonium bromide, benzyl-triethylammonium bromide, tetraethylammonium bromide and cetylpyridinium bromide. The
Neng-Chou Shang et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 37(2), 261-271 (2002-02-16)
This study investigates the variation of toxicity during ozonation of 2-chlorophenol (2-CP), 3-chlorophenol (3-CP) and 4-chlorophenol (4-CP) in neutral condition. Acute toxicity of pure chlorophenols (CPs) and their ozonated intermediates was evaluated by Microtox assay. The results revealed that the
Hideo Utsumi et al.
Water research, 37(20), 4924-4928 (2003-11-08)
Hydroxyl (OH) radical is proposed as an important factor in the ozonation of water. In the present study, the enhancing effect of 3-chlorophenol on OH radical generation was mathematically evaluated using electron spin resonance (ESR)/spin-trapping technique. OH radical was trapped
Stefano M Marino et al.
Archives of biochemistry and biophysics, 505(1), 67-74 (2010-09-30)
Tyrosinase (Ty) is a copper-containing enzyme ubiquitously distributed in nature. In recent years, Ty has attracted interest as a potential detoxifying agent for xenobiotic compounds with phenolic structure. Among these, chlorophenols are particularly relevant pollutants, commonly found in waste waters.
Clarissa A Olivati et al.
Bioprocess and biosystems engineering, 32(1), 41-46 (2008-04-15)
Langmuir-Blodgett (LB) and layer-by-layer films (LbL) of a PPV (p-phenylenevinylene) derivative, an azo compound and tetrasulfonated phthalocyanines were successfully employed as transducers in an "electronic tongue" system for detecting trace levels of phenolic compounds in water. The choice of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service