Skip to Content
Merck
All Photos(1)

Key Documents

34855-M

Sigma-Aldrich

Cyclohexane

suitable for HPLC, ≥99.7%

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€124.00
2.5 L
€276.00

€124.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 L
€124.00
2.5 L
€276.00

About This Item

Empirical Formula (Hill Notation):
C6H12
CAS Number:
Molecular Weight:
84.16
Beilstein:
1900225
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.21
grade:
HPLC grade
technique(s):
HPLC: suitable
bp:
80.7 °C (lit.)
vapor pressure:
168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)

€124.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

HPLC grade

vapor density

2.9 (vs air)

vapor pressure

168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)

Assay

≥99.7%

form

liquid

autoignition temp.

500 °F

expl. lim.

9 %

technique(s)

HPLC: suitable

impurities

≤0.0005% non-volatile matter
≤0.001% free acid (as HCl)
≤0.01% water (Karl Fischer)
≤1 ppb fluorescence (quinine) at 254 nm
≤1 ppb fluorescence (quinine) at 365 nm

evapn. residue

≤0.0005%

transmittance

220 nm, ≥20%
230 nm, ≥50%
235 nm, ≥80%
255 nm, ≥98%

refractive index

n20/D 1.426 (lit.)

bp

80.7 °C (lit.)

mp

4-7 °C (lit.)

density

0.779 g/mL at 25 °C (lit.)
0.780 g/mL at 25 °C

λ

H2O reference

UV absorption

λ: 210 nm Amax: ≤1.00
λ: 220 nm Amax: ≤0.50
λ: 230 nm Amax: ≤0.20
λ: 235 nm Amax: ≤0.10
λ: 240 nm Amax: ≤0.08
λ: 250 nm Amax: ≤0.03
λ: 255 nm Amax: ≤0.01

SMILES string

C1CCCCC1

InChI

1S/C6H12/c1-2-4-6-5-3-1/h1-6H2

InChI key

XDTMQSROBMDMFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Important notice
  • The article number 34855-4X2.5L-M will be discontinued. Please order the single bottle 34855-2.5L-M which is physically identical with the same exact specifications.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18.00 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service