Skip to Content
Merck
All Photos(1)

Documents

32994

Supelco

Thiosultap disodium

PESTANAL®, analytical standard

Synonym(s):

Disodium S,S′-[2-(dimethylamino)trimethylene]di(thiosulfate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11NNa2O6S4
CAS Number:
Molecular Weight:
355.38
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

[Na+].[Na+].CN(C)C(CSS([O-])(=O)=O)CSS([O-])(=O)=O

InChI

1S/C5H13NO6S4.2Na/c1-6(2)5(3-13-15(7,8)9)4-14-16(10,11)12;;/h5H,3-4H2,1-2H3,(H,7,8,9)(H,10,11,12);;/q;2*+1/p-2

InChI key

QSOHVSNIQHGFJU-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Antidotal effect of sodium dimercapto-propane sulphonate in the treatment of acute poisoning of 2-dimethylamino-1,3-histhio-sulfopropanedisodium salt in 20 cases].
Y F Di
Zhonghua nei ke za zhi, 32(12), 827-827 (1993-12-01)
Hui Chen et al.
Chemosphere, 84(5), 664-670 (2011-05-31)
Dunaliella salina, a unicellular green alga of environmental tolerance, was employed as test organism to investigate the toxicity effects of trichlorfon and dimehypo widely used in agriculture and veterinary as pesticides. The influences of trichlorfon and dimehypo on cell growth
[Antidotal effect of sodium dimercaptosuccinate against acute poisoning by the monosodium salt of 2-dimethylamino-1,3-bisthiosulfo-propane].
Z K Chen et al.
Zhongguo yao li xue bao = Acta pharmacologica Sinica, 6(3), 204-207 (1985-09-01)
Z Q Lu et al.
Zhonghua yu fang yi xue za zhi [Chinese journal of preventive medicine], 26(4), 213-215 (1992-07-01)
In mice, DMPS (250 mg/kg, i.v.) combined with diazepam (1.25 mg/kg, i.p.) could increase LD50 of p. o. SCD 5.3 times. DMPS (62.5 mg/kg, i.v.) antagonized completely the respiratory depression and neuromuscular blockade caused by SCD(7.5 mg/kg, i.v.) in rabbits.
Fengzu Zhang et al.
Bulletin of environmental contamination and toxicology, 88(3), 362-367 (2011-10-29)
A modified method for the analysis of monosultap residue in rice plant and environment was developed and validated. Monosultap residue dynamics and final residues in supervised field trials at GAP conditions were studied. At fortification levels of 0.05, 0.5 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service