Skip to Content
Merck
All Photos(1)

Documents

32597

Sigma-Aldrich

Diallyl N,N-diisopropylphosphoramidite

technical, ~90% (GC)

Synonym(s):

N,N-Bis(1-methylethyl)phosphoramidous acid di-2-propenyl ester, Diallyloxy(diisopropylamino)phosphine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH]2NP(OCH2CH=CH2)2
CAS Number:
Molecular Weight:
245.30
Beilstein:
4383514
MDL number:
UNSPSC Code:
12352100

grade

technical

Assay

~90% (GC)

refractive index

n20/D 1.456 (lit.)
n20/D 1.456

bp

130 °C (lit.)

density

0.928 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)N(C(C)C)P(OCC=C)OCC=C

InChI

1S/C12H24NO2P/c1-7-9-14-16(15-10-8-2)13(11(3)4)12(5)6/h7-8,11-12H,1-2,9-10H2,3-6H3

InChI key

QBLCHHSGJTUNSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Precursor involved in the synthesis of pharmacologically active molecules including:
  • Selective orally active S1P1 agonists
  • Water-soluble prodrugs of triazole CS-758 with antifungal activity
  • Fostriecin analogs as antitumor agents

Reactant involved in:
  • Phosphitylation of alcohols
  • Deallylation for the synthesis of nucleoside phosphoramidite
  • Posphitylation and stereoselective Pudovik rearrangement

Other Notes

Reagent used for the phosphorylation of alcohols

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W. Bannwarth et al.
Tetrahedron Letters, 30, 4219-4219 (1989)
J. Chen et al.
Tetrahedron Letters, 38, 969-969 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service