Skip to Content
Merck
All Photos(1)

Key Documents

31734

Supelco

β-Estradiol hemihydrate

VETRANAL®, analytical standard

Synonym(s):

1,3,5-Estratriene-3,17β-diol, 3,17β-Dihydroxy-1,3,5(10)-estratriene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24O2 · 0.5H2O
CAS Number:
Molecular Weight:
281.39
Beilstein:
1914275
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

O.C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1CC[C@@H]2O.C[C@]56CC[C@H]7[C@@H](CCc8cc(O)ccc78)[C@@H]5CC[C@@H]6O

InChI

1S/2C18H24O2.H2O/c2*1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20;/h2*3,5,10,14-17,19-20H,2,4,6-9H2,1H3;1H2/t2*14-,15-,16+,17+,18+;/m11./s1

InChI key

ZVVGLAMWAQMPDR-WVEWYJOQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jen-Liang Su et al.
The Journal of biological chemistry, 282(27), 19385-19398 (2007-05-22)
Osteoporosis is a major public health problem and the most obvious preventive strategy, hormone replacement therapy, has lost favor due to recent findings of the Women's Health Initiative regarding increased risks of breast cancer and cardiovascular disease. Resveratrol, a naturally
Artem Cherkasov et al.
Journal of medicinal chemistry, 49(25), 7466-7478 (2006-12-08)
A combination of protein-ligand docking and ligand-based QSAR approaches has been elaborated, aiming to speed-up the process of virtual screening. In particular, this approach utilizes docking scores generated for already processed compounds to build predictive QSAR models that, in turn
Pascal Pigeon et al.
Journal of medicinal chemistry, 48(8), 2814-2821 (2005-04-15)
A series of ruthenocene derivatives, 1-[4-(O(CH(2))(n)()N(CH(3))(2))phenyl]-1-(4-hydroxyphenyl)-2-ruthenocenylbut-1-ene, with n = 2-5, based on the structure of the breast cancer drug tamoxifen has been prepared. These compounds were obtained, via a McMurry cross-coupling reaction, as a mixture of Z and E isomers
Tomoko Soga et al.
Endocrinology, 155(5), 1944-1955 (2014-03-13)
Gonadotropin-inhibitory hormone (GnIH) neurons project to GnRH neurons to negatively regulate reproductive function. To fully explore the projections of the GnIH neurons, we created transgenic rats carrying an enhanced green fluorescent protein (EGFP) tagged to the GnIH promoter. With these
Steffen Renner et al.
Nature chemical biology, 5(8), 585-592 (2009-06-30)
The structure- and chemistry-based hierarchical organization of library scaffolds in tree-like arrangements provides a valid, intuitive means to map and navigate chemical space. We demonstrate that scaffold trees built using bioactivity as the key selection criterion for structural simplification during

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service