Skip to Content
Merck
All Photos(1)

Key Documents

31445-M

Sigma-Aldrich

Sodium peroxide

puriss. p.a., ACS reagent, reag. ISO, ≥95%

Synonym(s):

Sodium superoxide

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€395.00

€395.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 G
€395.00

About This Item

Empirical Formula (Hill Notation):
Na2O2
CAS Number:
Molecular Weight:
77.98
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.21
form:
powder

€395.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

ACS reagent
puriss. p.a.

Agency

reag. ISO

Assay

≥95%

form

powder

impurities

≤0.002% heavy metals (as Pb)
≤0.003% total nitrogen (N)

pH

12.8 (20 °C, 100 g/L)

mp

460 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤0.002%
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.001%

cation traces

Al: ≤0.001%
Fe: ≤0.002%

SMILES string

[Na+].[Na+].[O-][O-]

InChI

1S/2Na.O2/c;;1-2/q2*+1;-2

InChI key

PFUVRDFDKPNGAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium peroxide (Sodium superoxide) has been synthesized by the matrix reaction of sodium atoms and oxygen molecules at high dilution in argon. Its IR spectral studies have been reported.[1]

Application

Sodium peroxide may be used for the isotope dilution inductively coupled plasma mass spectrometric quantification of Ru, Pd, Ir, Pt and rare earth elements (such as Y, Th, Zr, Hf, Nb and Ta) in various geological samples.[2][3]

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Ox. Sol. 1 - Skin Corr. 1A

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service