Skip to Content
Merck
All Photos(1)

Documents

29285

Sigma-Aldrich

(R)-(−)-1-Cyclohexylethylamine

purum, ≥98.0% (sum of enantiomers, GC)

Synonym(s):

(R)-(−)-α-Methylcyclohexanemethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11CH(CH3)NH2
CAS Number:
Molecular Weight:
127.23
Beilstein:
2935070
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

grade

purum

Assay

≥98.0% (sum of enantiomers, GC)

optical activity

[α]20/D -3.8±0.3°, neat

optical purity

enantiomeric ratio: ≥99.0:1.0

refractive index

n20/D 1.463

bp

177-178 °C (lit.)

density

0.866 g/mL at 20 °C (lit.)

SMILES string

C[C@@H](N)C1CCCCC1

InChI

1S/C8H17N/c1-7(9)8-5-3-2-4-6-8/h7-8H,2-6,9H2,1H3/t7-/m1/s1

InChI key

XBWOPGDJMAJJDG-SSDOTTSWSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Chiral auxiliary; used for preparing chiral polymethacrylamides, which are employed as stationary phases for the liquid-chromatographic resolution of racemic amides ; Preparation of polyamides bound to silica gel for HPLC which are pressure-stable

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G. Blaschke et al.
Angewandte Chemie (International Edition in English), 98, 808-808 (1986)
G. Blaschke
Journal of Liquid Chromatography, 9, 341-341 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service