Skip to Content
Merck
All Photos(1)

Documents

288519

Supelco

Silica gel

high-purity grade, pore size 60 Å, 5-25 μm particle size, without binder, suitable for thin layer chromatography (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41115711
PubChem Substance ID:
NACRES:
NB.21

material

plastic support
silica gel 60 matrix

Quality Level

grade

high-purity grade

form

powder

feature

binder Organic Polymer
fluorescent indicator: no

packaging

pkg of 250 g

manufacturer/tradename

Sigma-Aldrich

technique(s)

thin layer chromatography (TLC): suitable

surface area

~500 m2/g

impurities

≤0.001% Fe3+

particle size

5-25 μm

pore size

0.75 cm3/g pore volume
60 Å medium pore diameter
60 Å pore size

pH

~6.8 (10% in aq. suspension)

bp

2230 °C

mp

>1600 °C

anion traces

chloride (Cl-): ≤0.003%

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silica gel is a rigid 3D network of colloidal silica. It is exclusively used as a support material for the active titanium(III) centers in Ziegler-Natta catalysts. It allows a controlled fragmentation resulting in the formation of uniform polymer particles with narrow particle size distribution and high bulk density. Silica gel is classified as below:
a. Aquagel - pores are filled with water
b. Xerogel - by the process of evaporation, aqueous phase in the pores are removed
c. Aerogel - solvent removed by supercritical extraction

Application

Silica gel was used on glass plates to perform preparative thin layer chromatography.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

F. Daayf et al.
Plant physiology, 113(3), 719-727 (1997-03-01)
Phenolic compounds extracted from cucumber (Cucumis sativus L.) leaves were separated and analyzed for their differential presence and fungitoxicity in relation to a prophylactic treatment with Milsana (Compo, Munster, Germany) against powdery mildew (Sphaerotheca fuliginea). Based on our extraction and
A simple method for production of pure silica from rice hull ash
Kalapathy, U., A. Proctor, and J. Shultz.
Bioresource Technology, 73, 257-262 (2000)
Silica gel supported zirconocene dichloride/methylalumoxane catalysts for ethylene polymerization: Effects of heterogenation on activity, polymer microstructure and product morphology.
Janiak, Christoph, and Bernhard Rieger.
Angew. Makromol. Chem., 215, 47-57 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service