235482
Chlorophenol Red sodium salt
Dye content 70 %
Synonym(s):
3′,3′′-Dichlorophenolsulfonephthalein sodium salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C19H11Cl2NaO5S
CAS Number:
Molecular Weight:
445.25
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
composition
Dye content, 70%
λmax
575 nm
SMILES string
[Na+].Oc1ccc(cc1Cl)C(=C2\C=CC(=O)C(Cl)=C2)\c3ccccc3S([O-])(=O)=O
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service