Skip to Content
Merck
All Photos(1)

Documents

18600

Sigma-Aldrich

11-Bromo-1-undecanol

purum, ≥99.0% (AT)

Synonym(s):

Undecamethylene bromohydrin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)11OH
CAS Number:
Molecular Weight:
251.20
Beilstein:
1741180
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Quality Level

Assay

≥99.0% (AT)

form

solid

bp

165-170 °C/1 mmHg (lit.)

mp

45-48 °C
46-49 °C (lit.)

solubility

chloroform: soluble 50 mg/mL, clear to slightly hazy, colorless to dark yellow

SMILES string

OCCCCCCCCCCCBr

InChI

1S/C11H23BrO/c12-10-8-6-4-2-1-3-5-7-9-11-13/h13H,1-11H2

InChI key

XFGANBYCJWQYBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

11-Bromo-1-undecanol was used in the preparation of 11-mercapto-1-undecanol and 1-(2-acryloyloxyundecyl)-3-methylimidazolium bromide. It was also used in one-pot synthesis of graft copolymers based on a metathesis skeleton with poly(methyl methacrylate) grafts.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feng Yan et al.
Chemical communications (Cambridge, England), (25)(25), 2696-2698 (2006-06-21)
Surfactants based on imidazolium ionic liquids (ILs), including polymerizable surfactant ILs, have been synthesized and used to stabilize polymerizable microemulsions useful for producing polymer nanoparticles, gels, and open-cell porous materials.
Vibrational spectroscopic studies of the attachment chemistry for zirconium phosphonate multilayers at gold and germanium surfaces.
Frey BL, et al
Langmuir, 9(7), 1815-1820 (1993)
One-pot, one-catalyst synthesis of graft copolymers by controlled ROMP and ATRP polymerizations.
Charvet R and Novak BM.
Macromolecules, 37(23), 8808-8811 (2004)
Ceren Kutlu Kaya et al.
Journal of biomedical materials research. Part A, 108(3), 760-759 (2019-12-04)
Hypothyroidism is an autoimmune disease associated with underactive thyroid gland. In this study, a dual effect polymeric system was designed to release Cepharanthine (CEP) to block T cell activation and Selenium (Se) to decrease the anti-thyroid peroxidase (TPOAb) concentration in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service