Skip to Content
Merck
All Photos(1)

Key Documents

17205

Millipore

Potato Glucose Sucrose Agar

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171605
NACRES:
NA.85

form

powder

Quality Level

shelf life

limited shelf life, expiry date on the label

composition

agar, 15 g/L
dextrose (glucose), 40 g/L
potatoes, infusion from (solids), 4 g/L
sucrose, 600 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

application(s)

food and beverages
microbiology

suitability

nonselective for Zygosaccharomyces spp.

Application

For the isolation and cultivation of Zygosaccharomyces rouxii from chocolate syrup.

Preparation Note

Suspend 65.9 g in 100 ml distilled water. Heat to dissolve the medium with stirring. Sterilize by autoclaving at 121°C for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ramkumar Menon et al.
Aging, 8(2), 216-230 (2016-02-07)
Aging is associated with the onset of several diseases in various organ systems; however, different tissues may age differently, rendering some of them dysfunctional sooner than others. Placental membranes (fetal amniochorionic membranes) protect the fetus throughout pregnancy, but their longevity

Articles

Yeast is one of the most important microorganisms known and utilised by mankind. Ancient Middle Eastern civilisations used the organism to bake bread and to produce mead, beer and wine.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service