Skip to Content
Merck
All Photos(1)

Key Documents

16898

Supelco

Hypaconitine

analytical standard

Synonym(s):

16,16-trimethoxy-4-(methoxymethyl)-20-methyl-,8-acetate benzoate,(1-α,6-α,14-α,15-α,16-β)-aconitane-8,13,14,15-tetraol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H45NO10
CAS Number:
Molecular Weight:
615.71
UNSPSC Code:
12352200
NACRES:
NA.24

grade

analytical standard

Assay

≥97.5% (HPLC)

optical activity

[α]/D 22.5±2.5°, c = 0.1 in chloroform

shelf life

limited shelf life, expiry date on the label

format

neat

storage temp.

2-8°C

SMILES string

O=C(C1=CC=CC=C1)O[C@H]2[C@@]3(O)[C@@H](OC)[C@H](O)[C@]4(OC(C)=O)C5[C@H](OC)C6C([C@@H](OC)CC[C@]6(CN7C)COC)(C75)[C@H](C3)C42

InChI

1S/C33H45NO10/c1-17(35)44-33-21-19(14-31(38,28(42-6)26(33)36)27(21)43-29(37)18-10-8-7-9-11-18)32-20(40-4)12-13-30(16-39-3)15-34(2)25(32)22(33)23(41-5)24(30)32/h7-11,19-28,36,38H,12-16H2,1-6H3/t19-,20+,21?,22?,23+,24?,25?,26+,27-,28+,30+,31-,32?,33-/m1/s1

InChI key

FIDOCHXHMJHKRW-XCIBRDTCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hypaconitine is a constituent of diester-diterpene Aconitum alkaloids (AAs), which are found in aconite plants.

Application

Hypaconitine may be used as an analytical standard for the determination of the analyte in biological samples, Aconitum tubers by liquid chromatography (LC) based techniques.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of aconitine, hypaconitine and mesaconitine in urine using hollow fiber liquid-phase microextraction combined with high-performance liquid chromatography
Yang Y, et al.
Journal of Chromatography. B, Biomedical Applications, 878(28), 2811-2816 (2010)
Simultaneous determination of aconitine, mesaconitine, hypaconitine, bulleyaconitine and lappaconitine in human urine by liquid chromatography-electrospray ionization-tandem mass spectrometry
Li M and Cai Z
Analytical Methods : Advancing Methods and Applications, 5(16), 4034-4038 (2013)
Rina Kaneko et al.
Journal of mass spectrometry : JMS, 41(6), 810-814 (2006-06-14)
The Aconitum species (Ranunculaceae) are widely distributed in northern Asia and North America. Their roots are popularly used in herbal medicines in China and Japan. Many cases of accidental, suicidal and homicidal intoxication with this plant have been reported; some
Ying Xie et al.
Journal of chromatography. A, 1093(1-2), 195-203 (2005-10-20)
By optimizing the extraction, separation and analytical conditions, a reliable and accurate high-performance liquid chromatography (HPLC) method coupled with photodiode array detector (DAD) was developed for simultaneous quantitative determination of six Aconitum alkaloids, i.e., aconitine, mesaconitine, hypaconitine, benzoylaconine, benzoylmesaconine and
Lei-ping He et al.
Journal of analytical toxicology, 33(9), 588-594 (2009-12-31)
A rapid, sensitive, and specific liquid chromatography-electrospray ionization-tandem mass spectrometry (LC-ESI-MS-MS) method was developed and validated for simultaneous determination of aconitine (AC), mesaconitine (MA), and hypaconitine (HA), the three toxic constituents from Sini decoction (SND) in rat plasma. After the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service