Skip to Content
Merck
All Photos(1)

Documents

09784

Sigma-Aldrich

(1S,3R)-(+)-3-(Boc-amino)cyclopentanecarboxylic acid

≥96.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19NO4
CAS Number:
Molecular Weight:
229.27
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥96.0% (HPLC)

form

crystalline

optical activity

[α]/D +16.0±1°, c = 1% in methanol

reaction suitability

reaction type: Boc solid-phase peptide synthesis

color

beige

mp

110-114 °C

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N[C@@H]1CC[C@@H](C1)C(O)=O

InChI

1S/C11H19NO4/c1-11(2,3)16-10(15)12-8-5-4-7(6-8)9(13)14/h7-8H,4-6H2,1-3H3,(H,12,15)(H,13,14)/t7-,8+/m0/s1

InChI key

RNJQBGXOSAQQDG-JGVFFNPUSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bioluminescent assay of the adenylate energy charge.
S R Ford et al.
Methods in molecular biology (Clifton, N.J.), 102, 69-81 (1998-07-29)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service