Skip to Content
Merck
All Photos(2)

Documents

09655

Sigma-Aldrich

2-(3-Methylbutyryl)-5,5-dimethyl-1,3-cyclohexandione

≥99.0% (GC)

Synonym(s):

2-Isovaleryldimedone, 5,5-Dimethyl-2-(3-methylbutyryl)-1,3-cyclohexanedione, Ddiv

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20O3
CAS Number:
Molecular Weight:
224.30
Beilstein:
3275155
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (GC)

refractive index

n20/D 1.498

density

1.027 g/mL at 20 °C (lit.)

application(s)

peptide synthesis

SMILES string

CC(C)CC(=O)C1C(=O)CC(C)(C)CC1=O

InChI

1S/C13H20O3/c1-8(2)5-9(14)12-10(15)6-13(3,4)7-11(12)16/h8,12H,5-7H2,1-4H3

InChI key

HNPWTDUZIXAJSA-UHFFFAOYSA-N

Other Notes

Amine protecting group which is orthogonal to Fmoc, Boc and tert-butyl protections; selective cleavage with 2% hydrazine in DMF.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beatriz Reina Bouvet et al.
Archivos espanoles de urologia, 58(10), 1049-1054 (2006-02-18)
To investigate sperm quality before and after swim up in infertile patients, and to compare it with a fertile men population. Semen samples from 55 patients consulting at the infertility services of the Hospitals "Centenario" in Rosario and "Eva Perón"
S.R. Chhabra et al.
Tetrahedron Letters, 39, 1603-1603 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service