Skip to Content
Merck
All Photos(1)

Key Documents

05689

Supelco

Diethyl ethoxymethylenemalonate

for LC-MS derivatization, LiChropur, ≥97.0% (GC)

Synonym(s):

DEEMM, Ethoxymethylenemalonic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCH=C(COOC2H5)2
CAS Number:
Molecular Weight:
216.23
Beilstein:
880058
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (GC)

form

liquid

quality

LiChropur

technique(s)

LC/MS: suitable

refractive index

n20/D 1.462 (lit.)

bp

279-281 °C (lit.)

density

1.07 g/mL at 25 °C (lit.)

SMILES string

CCO\C=C(\C(=O)OCC)C(=O)OCC

InChI

1S/C10H16O5/c1-4-13-7-8(9(11)14-5-2)10(12)15-6-3/h7H,4-6H2,1-3H3

InChI key

LTMHNWPUDSTBKD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

identification (LC-MS): identity conforms

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1A

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

291.2 °F - closed cup

Flash Point(C)

144 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Riin Rebane et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 904, 99-106 (2012-08-15)
Amino acid analysis with high performance liquid chromatography with electrospray ionization mass spectrometry (LC-ESI-MS) is an emerging method. For more sensitive analysis, derivatization is used and next to commercially available derivatization reagents such as dansyl chloride (DNS), 9-fluorenylmethyl chloroformate (FMOC-Cl)
Marta Perez et al.
Applied microbiology and biotechnology, 99(8), 3547-3558 (2014-12-23)
Enterococcus faecalis is a commensal bacterium of the human gut that requires the ability to pass through the stomach and therefore cope with low pH. E. faecalis has also been identified as one of the major tyramine producers in fermented
Cristina Megías et al.
Talanta, 131, 95-98 (2014-10-05)
A method for determination of the non-protein amino acid l-α-amino-γ-(guanidinooxy)-n-butyric acid (L-canavanine) and other free amino acids in Vicia disperma is presented. Seed extracts were derivatized by reaction with diethyl ethoxymethylenemalonate and analyzed by reverse-phase high-performance liquid chromatography. Calibration curves
J L Ordóñez et al.
Food chemistry, 178, 221-228 (2015-02-24)
This paper studies the amino acid profile of beverages obtained through the fermentation of strawberry purée by a surface culture using three strains belonging to different acetic acid bacteria species (one of Gluconobacter japonicus, one of Gluconobacter oxydans and one
Maarja-Liisa Oldekop et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 967, 147-155 (2014-08-12)
For liquid chromatographic analysis of amino acids involving derivatization and mass-spectrometric detection, it becomes more important to evaluate the presence of matrix effects in complex samples. This is somewhat complicated for amino acid analysis where analyte free sample matrix is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service