Skip to Content
Merck
All Photos(1)

Key Documents

03800

Sigma-Aldrich

Ethylene glycol diglycidyl ether

technical, ~50% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O4
CAS Number:
Molecular Weight:
174.19
Beilstein:
110424
EC Number:
MDL number:
UNSPSC Code:
12162002
Pricing and availability is not currently available.

grade

technical

concentration

~50% (GC)

refractive index

n20/D 1.463

bp

112 °C/4.5 mmHg (lit.)

density

1.118 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C(COCC1CO1)OCC2CO2

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service