Skip to Content
Merck
All Photos(1)

Key Documents

00456

Supelco

2-{2-[(2-Bromo-6-chlorophenyl)amino]phenyl}acetic acid

pharmaceutical impurity standard

Synonym(s):

Diclofenac impurity D (PhEur), 2-[(2-Bromo-6-chlorophenyl)amino]benzeneacetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€441.00

€441.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€441.00

About This Item

Empirical Formula (Hill Notation):
C14H11BrClNO2
CAS Number:
Molecular Weight:
340.60
Beilstein:
4195335
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€441.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98.0% (HPLC)

form

powder

shelf life

limited shelf life, expiry date on the label

impurities

≤0.5% water

application(s)

clinical testing

InChI

1S/C14H11BrClNO2/c15-10-5-3-6-11(16)14(10)17-12-7-2-1-4-9(12)8-13(18)19/h1-7,17H,8H2,(H,18,19)

InChI key

RABKUCPRNJPBRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Repr. 2 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katarzyna Rolf et al.
Biological trace element research, 199(8), 3013-3020 (2020-10-19)
Deficient human diet is usually reduced in many nutrients, but animal studies on iron absorption have been only carried out for rats fed well-balanced (control) and iron-deficient diets. The aim of this study was to evaluate the effect of iron

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service