Skip to Content
Merck
All Photos(1)

Key Documents

24-5310

Sigma-Aldrich

Potassium phosphate tribasic hydrate

SAJ first grade, ≥97.0% dry basis

Synonym(s):

Tripotassium phosphate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K3PO4
Molecular Weight:
212.27 (anhydrous basis)
UNSPSC Code:
12352302
grade:
SAJ first grade
form:
powder
Pricing and availability is not currently available.

grade

SAJ first grade

Assay

≥97.0% dry basis

form

powder

availability

available only in Japan

pH

12 (20 °C, 10 g/L)

storage temp.

15-25°C

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service