Skip to Content
Merck
All Photos(1)

Key Documents

19-0510

Sigma-Aldrich

Magnesium sulfate

SAJ first grade (dried), ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MgSO4
CAS Number:
Molecular Weight:
120.37
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
SAJ first grade (dried)
form:
solid
Pricing and availability is not currently available.

grade

SAJ first grade (dried)

vapor density

<0.01 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

≥99.0%

form

solid

availability

available only in Japan

pH

~7.9 (25 °C, 50 g/L)

SMILES string

[Mg++].[O-]S([O-])(=O)=O

InChI

1S/Mg.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

CSNNHWWHGAXBCP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lex W Doyle et al.
Obstetrics and gynecology, 113(6), 1327-1333 (2009-05-23)
To systematically review rates of neurologic outcomes reported in childhood for the preterm fetus exposed to antenatal magnesium sulfate. We searched the Cochrane Pregnancy and Childbirth Group's Trials Register, CENTRAL (The Cochrane Library 2008, Issue 3), relevant references from retrieved
Lex W Doyle
Current opinion in pediatrics, 24(2), 154-159 (2012-01-10)
Antenatal magnesium sulfate may reduce the excessive rates of cerebral palsy in survivors of very preterm birth. There are five randomized controlled trials of magnesium sulfate therapy given to the mother prior to very preterm birth which have reported neurological
Claire L O'Brien et al.
Gut, 63(10), 1596-1606 (2014-01-17)
Our aim was to determine whether or not specific microorganisms were transported selectively to lymph nodes in Crohn's disease (CD) by comparing node and mucosal microbial communities in patients and controls. We also sought evidence of dysbiosis and bacterial translocation.
Federica Belluti et al.
ChemMedChem, 9(2), 371-382 (2014-01-10)
A significant improvement in the treatment of trypanosomiases has been achieved with the recent development of nifurtimox-eflornithine combination therapy (NECT). As an alternative to drug combinations and as a means to overcome most of the antitrypanosomatid drug discovery challenges, a
Robert J Floor et al.
Chembiochem : a European journal of chemical biology, 15(11), 1660-1672 (2014-07-01)
We explored the use of a computational design framework for the stabilization of the haloalkane dehalogenase LinB. Energy calculations, disulfide bond design, molecular dynamics simulations, and rational inspection of mutant structures predicted many stabilizing mutations. Screening of these in small

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service