Skip to Content
Merck
All Photos(1)

Documents

18-1010

Sigma-Aldrich

L-Leucine

SAJ special grade, ≥99.0%

Synonym(s):

(S)-2-Amino-4-methylpentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
131.17
Beilstein:
1721722
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

grade

SAJ special grade

Assay

≥99.0%

availability

available only in Japan

mp

>300 °C (lit.)

application(s)

peptide synthesis

SMILES string

CC(C)C[C@H](N)C(O)=O

InChI

1S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t5-/m0/s1

InChI key

ROHFNLRQFUQHCH-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sebastian Kalamajski et al.
Matrix biology : journal of the International Society for Matrix Biology, 29(4), 248-253 (2010-01-19)
Small leucine-rich proteoglycans/proteins (SLRPs) are associated with collagen fibril formation, and therefore important for the proper formation of extracellular matrices. SLRPs are differentially expressed in tissues and during pathological conditions, contributing to the development of connective tissue properties. The binding
Qian Wang et al.
Journal of the National Cancer Institute, 105(19), 1463-1473 (2013-09-21)
L-type amino acid transporters (LATs) uptake neutral amino acids including L-leucine into cells, stimulating mammalian target of rapamycin complex 1 signaling and protein synthesis. LAT1 and LAT3 are overexpressed at different stages of prostate cancer, and they are responsible for
M Sakamoto et al.
The Journal of clinical endocrinology and metabolism, 56(1), 202-204 (1983-01-01)
Recent studies on the nucleotide sequence of cloned DNA complementary to mRNA for preproenkephalin from bovine adrenal medulla and human pheochromocytoma have revealed that this precursor molecule contains four copies of methionine-enkephalin (Met-Enk) and one copy each of leucine-enkephalin (Leu-Enk)
Gaetano Santulli et al.
The Journal of clinical investigation, 125(5), 1968-1978 (2015-04-07)
The type 2 ryanodine receptor (RyR2) is a Ca2+ release channel on the endoplasmic reticulum (ER) of several types of cells, including cardiomyocytes and pancreatic β cells. In cardiomyocytes, RyR2-dependent Ca2+ release is critical for excitation-contraction coupling; however, a functional
T Yoshimasa et al.
The Journal of clinical investigation, 69(3), 643-650 (1982-03-01)
To elucidate the physiological and pathophysiological significance of methionine- and leucine-enkephalin (Met-and Leu-enkephalin, respectively) in human sympathoadrenal system, the contents of these peptides in normal human sympathetic nervous system, adrenal medulla, and pheochromocytomas were determined by specific radioimmunoassays combined with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service