Skip to Content
Merck
All Photos(1)

Key Documents

18-0130

Sigma-Aldrich

D-Lactose monohydrate

SAJ first grade

Synonym(s):

β-D-Gal-(1→4)-D-Glc, 4-O-β-D-Galactopyranosyl-D-glucose, Milk sugar

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11 · H2O
CAS Number:
Molecular Weight:
360.31
Beilstein:
3768231
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ first grade

form

solid

availability

available only in Japan

SMILES string

O.OC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C=O

InChI

1S/C12H22O11.H2O/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12;/h1,4-12,14-21H,2-3H2;1H2/t4-,5+,6+,7+,8-,9-,10+,11+,12-;/m0./s1

InChI key

HBDJFVFTHLOSDW-XBLONOLSSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fan Zhou et al.
Scientific reports, 1, 78-78 (2012-02-23)
The invisibility cloak has been a long-standing dream for many researchers over the decades. Using transformation optics, a three-dimensional (3D) object is perceived as having a reduced number of dimensions, making it "undetectable" judging from the scattered field12345. Despite successful
Akhtar Siddiqui et al.
Journal of pharmaceutical sciences, 103(9), 2819-2828 (2014-03-04)
The objective of this study was to develop powder X-ray diffraction (XRPD) chemometric model for quantifying crystalline tacrolimus from solid dispersion (SD). Three SDs (amorphous tacrolimus component) with varying drug to excipient ratios (24.4%, 6.7%, and 4.3% drug) were prepared.
J Nijdam et al.
Colloids and surfaces. B, Biointerfaces, 123, 53-60 (2014-09-30)
Segregation of the protein bovine serum albumin (BSA) and lactose in thin aqueous films during drying was investigated by examining the composition of the dried films using inverse micro Raman spectroscopy (IMRS) and X-ray photoelectron spectroscopy (XPS) sputter-depth profiling. The
Daniel Cavanagh et al.
Applied and environmental microbiology, 81(12), 3961-3972 (2015-04-05)
Lactococcus lactis is predominantly associated with dairy fermentations, but evidence suggests that the domesticated organism originated from a plant niche. L. lactis possesses an unusual taxonomic structure whereby strain phenotypes and genotypes often do not correlate, which in turn has
Akhtar Siddiqui et al.
International journal of pharmaceutics, 493(1-2), 1-6 (2015-07-26)
The objective of this study was to develop and validate XRPD analytical method for the estimation of percent crystalline warfarin sodium present in drug products. Warfarin sodium (WS) is a clathrate containing Isopropyl alcohol entrapped in the crystalline structure. Four

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service