Skip to Content
Merck
All Photos(1)

Key Documents

15-0510

Sigma-Aldrich

myo-Inositol

SAJ special grade, ≥99.0%

Synonym(s):

1,2,3,4,5,6-Hexahydroxycyclohexane, i-Inositol, meso-Inositol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1907329
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ special grade

vapor density

6.2 (vs air)

Assay

≥99.0%

form

solid

availability

available only in Japan

mp

222-227 °C (lit.)

SMILES string

O[C@H]1[C@H](O)[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-12H/t1-,2-,3-,4+,5-,6-

InChI key

CDAISMWEOUEBRE-GPIVLXJGSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

A component of membrane phospholipids, glycosyl­phosphatidyl­inositol anchors that bind glycoproteins to cell membranes, and inositol phosphate second messengers.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Hallman et al.
Early human development, 10(3-4), 245-254 (1985-01-01)
According to animal studies myoinositol decreases surfactant phosphatidylglycerol and increases phosphatidylinositol. In the present study lung effluent phospholipids and serum myoinositol were analyzed in respiratory distress syndrome (RDS, 19 cases), in other lung disease (6 cases) and in 22 newborn
M Utriainen et al.
Journal of neuro-oncology, 62(3), 329-338 (2003-06-05)
The signal of choline containing compounds (Cho) in proton magnetic resonance spectroscopy (1H-MRS) is elevated in brain tumors. [11C]choline uptake as assessed using positron emission tomography (PET) has also been suggested to be higher in brain tumors than in the
Pascal M Groenen et al.
American journal of obstetrics and gynecology, 189(6), 1713-1719 (2004-01-08)
The purpose of this study was to investigate the maternal and children's myo-inositol, glucose, and zinc status in association with spina bifida risk. Sixty-three mothers and 70 children with spina bifida and 102 control mothers and 85 control children were
C Servo et al.
Diabetologia, 11(6), 575-580 (1975-12-01)
Cerebrospinal fluid (CSF) or CSF and plasma levels of sorbitol, 1,5-anhydroglucitol and myoinositol of diabetic and non-diabetic patients with normal kidney function and of diabetic and non-diabetic patients with impaired kidney function were measured by gas-liquid chromatography. The CSF sorbitol
L Chang et al.
Neurology, 53(4), 782-789 (1999-09-17)
To determine whether cerebral metabolite abnormalities normalize with highly active antiretroviral therapy (HAART). Patients with HIV-cognitive motor complex (HIV-CMC) show cerebral metabolite abnormalities in the early stages of dementia. Sixteen patients with HIV-CMC were evaluated before and after HAART, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service