Skip to Content
Merck
All Photos(1)

Key Documents

09-1840

Sigma-Aldrich

Diethyl ether

≥99.5%, suitable for residual phthalate analysis

Synonym(s):

Ether, Ethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)2O
CAS Number:
Molecular Weight:
74.12
Beilstein:
1696894
MDL number:
UNSPSC Code:
12000000

vapor density

2.6 (vs air)

Assay

≥99.5%

autoignition temp.

320 °F

expl. lim.

36.5 %

availability

available only in Japan

refractive index

n20/D 1.3530 (lit.)

bp

34.6 °C (lit.)

mp

−116 °C (lit.)

density

0.706 g/mL at 25 °C (lit.)

suitability

suitable for residual phthalate analysis

SMILES string

CCOCC

InChI

1S/C4H10O/c1-3-5-4-2/h3-4H2,1-2H3

InChI key

RTZKZFJDLAIYFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan E Camacho Londoño et al.
European heart journal, 36(33), 2257-2266 (2015-06-13)
Pathological cardiac hypertrophy is a major predictor for the development of cardiac diseases. It is associated with chronic neurohumoral stimulation and with altered cardiac Ca(2+) signalling in cardiomyocytes. TRPC proteins form agonist-induced cation channels, but their functional role for Ca(2+)
Tatsuya Yoshino et al.
Organic letters, 14(16), 4290-4292 (2012-08-09)
A highly efficient total synthesis of the 11-membered cyclic aspercyclides A (1) and B (2) has been achieved by chemo- and regioselective intramolecular oxidative C-O bond formation from differently substituted diphenols.
Faysal Benaskar et al.
ChemSusChem, 6(2), 353-366 (2012-11-30)
A μ(2)-process in the Ullmann-type C-O coupling of potassium phenolate and 4-chloropyridine was successfully performed in a combined microwave (MW) and microflow process. Selective MW absorption in a micro-fixed-bed reactor (μ-FBR) by using a supported Cu nanocatalyst resulted in an
Ether-directed ortho-C-H olefination with a palladium(II)/monoprotected amino acid catalyst.
Gang Li et al.
Angewandte Chemie (International ed. in English), 52(4), 1245-1247 (2012-12-15)
Julia J Griese et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(43), 17189-17194 (2013-10-09)
Although metallocofactors are ubiquitous in enzyme catalysis, how metal binding specificity arises remains poorly understood, especially in the case of metals with similar primary ligand preferences such as manganese and iron. The biochemical selection of manganese over iron presents a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service