Skip to Content
Merck
All Photos(1)

Documents

05-0400

Sigma-Aldrich

Calcium acetate hydrate

JIS special grade, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Ca · xH2O
CAS Number:
Molecular Weight:
158.17 (anhydrous basis)
Beilstein:
3692527
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

JIS special grade

Assay

≥99.0%

availability

available only in Japan

SMILES string

O.CC(=O)O[Ca]OC(C)=O

InChI

1S/2C2H4O2.Ca.H2O/c2*1-2(3)4;;/h2*1H3,(H,3,4);;1H2/q;;+2;/p-2

InChI key

XQKKWWCELHKGKB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dilek Teker et al.
Materials science & engineering. C, Materials for biological applications, 48, 579-585 (2015-01-13)
An innovative multi-layer coating comprising a bioactive compound layer (consisting of hydroxyapatite and calcium titanate) with an underlying titanium oxide layer (in the form of anatase and rutile) has been developed on Grade 4 quality commercially pure titanium via a
Hamid Esfahani et al.
Journal of colloid and interface science, 443, 143-152 (2014-12-30)
Surface modification of electrospun polymeric membrane surfaces is a critical step towards the separation process including protein adsorption. In this study, the electrospun Nylon fibers was incorporated with positively charged zinc doped hydroxyapatite (HAp) nanoparticles to study the adsorption of
S Yoon et al.
Cell death & disease, 5, e1494-e1494 (2014-10-31)
Apoptosis, which is anti-inflammatory, and necrosis, which is pro-inflammatory, represent the extremes of the cell death spectrum. Cell death is complex and both apoptosis and necrosis can be observed in the same cells or tissues. Here, we introduce a novel
Tanja Rozman Peterka et al.
Journal of pharmaceutical and biomedical analysis, 110, 67-75 (2015-03-26)
Tacrolimus is macrolide drug that is widely used as a potent immunosuppressant. In the present work compatibility testing was conducted on physical mixtures of tacrolimus with excipients and on compatibility mixtures prepared by the simulation of manufacturing process used for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service