Key Documents
ZWDKADPA1
Milli-Q
Washer Distribution Kit Adaptor
Connects a washer distribution kit to the outlet valve of the Milli-Q® IQ/IX storage tank
Select a Size
ZWDKADPA1
€178.00
€178.00
Please contact Customer Service for Availability
Request more information
Select a Size
Change View
ZWDKADPA1
€178.00
About This Item
UNSPSC Code:
41104214
NACRES:
JA.13
€178.00
Please contact Customer Service for Availability
Request more information
compatibility
for use with Milli-Q® IX 7003
for use with Milli-Q® IX 7005
for use with Milli-Q® IX 7010
for use with Milli-Q® IX 7015
for use with Milli-Q® IQ 7003
for use with Milli-Q® IQ 7005
for use with Milli-Q® IQ 7010
for use with Milli-Q® IQ 7015
General description
The Washer Distribution Kit Adaptor allows to connect a Washer Distribution Kit to the outlet valve below the Milli-Q® IQ/IX storage tank.
Features and Benefits
This connection allows to feed the Washer Distribution Kit with pure water from Milli-Q® IQ/IX storage tank.
Legal Information
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany