Skip to Content
Merck
All Photos(1)

Key Documents

ZLXLALCAB

Milli-Q

Alarm Relay Cable

For use with Milli-Q® high-flow water purification systems

Synonym(s):

Alarm cable-2x 24 vdc 4 w, 2x 4-20 mA

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€207.00

€207.00


Estimated to ship onApril 15, 2025Details


Request more information

Select a Size

Change View
1 EA
€207.00

About This Item

UNSPSC Code:
41104214
NACRES:
JA.13

€207.00


Estimated to ship onApril 15, 2025Details


Request more information

packaging

pkg of 1 unit

compatibility

for use with Milli-Q® CLX
for use with Milli-Q® HR
for use with Milli-Q® HX

Looking for similar products? Visit Product Comparison Guide

General description

This alarm output allows remote monitoring of alerts and alarms through an external device and/or through the MyMilli-Q remote service feature.

Application

For use with Milli-Q® CLX/HX/HXSD/HR water purification systems

Legal Information

Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
MyMilli-Q is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service