Skip to Content
Merck
All Photos(1)

Key Documents

ZLXEV05WW

Milli-Q

Elix® Essential 5 UV

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104202
NACRES:
SB.53

product line

Elix® Essential

Quality Level

Other Notes

Directions for Use
  • Organism Retention: Microorganisms
  • Mode of Action: Filtering and UV
  • Application: General laboratory analysis
  • Intended Use: Water purification.
  • Instructions for Use: This item provides water filtered through 0.22 μm sterilizing-grade membranes and exposed to bactericidal UV lamp. Please refer to system equipment user guide section "System" (provided on USB key in the North American configuration version).
  • Storage Statement: Store in dry location.
  • Disposal Statement: Dispose of in accordance with applicable federal, state, and local regulations.

Legal Information

ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service