Key Documents
XX4404704
Millipore
SS PTFE-Coated Back-Pressure Support
Select a Size
XX4404704
€167.00
€167.00
Please contact Customer Service for Availability
Select a Size
Change View
XX4404704
€167.00
About This Item
UNSPSC Code:
41104909
eCl@ss:
32031613
NACRES:
NB.24
€167.00
Please contact Customer Service for Availability
material
stainless steel
Quality Level
diam.
47 mm
General description
Applications:
In-Line Stainless Steel Filter Holders are commonly used for the filtration of gases or liquids at inlet pressures of 20 to 700 bar. Differential pressure depends on filter type used. All filter holders may be autoclaved (available in 25 mm and 47 mm).
Filter holder XX4404700 is equipped with a back-pressure support screen. It may be autoclaved with filter in-place, if PTFE-coated support screens, XX4404702 and XX4404704, are used.
- XX4204709, XX4304701, XX4404703, 5614, and 00907P are replacement parts for Filter Holder XX4404700.
Filter holder XX4504700 may be equipped with an upstream filter support screen, XX4504704, for back-pressure protection of filter.
- XX4504704, XX4504705, XX4504710, XX4504713, and XX45047RK are replacement parts for Filter Holder XX4504700.
Replacement parts for the 25mm SS Filter Holder are available as a kit under Catalog Number XX45025RK, which includes: (3) stainless steel hex-cap screws, (3) LCR-treated Buna-N O-rings (2-018), (1) stainless steel support screen, (1) Allen wrench.