Skip to Content
Merck

Skip To

TX1395

2,2,4-Trimethylpentane

Practical

Synonym(s):

Isooctane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
(CH3)2CHCH2C(CH3)3
CAS Number:
Molecular Weight:
114.23
EC Index Number:
208-759-1
UNSPSC Code:
12191502
Beilstein/REAXYS Number:
1696876
MDL number:
Assay:
≥95% (GC)
Bp:
98-99 °C (lit.)
Vapor pressure:
41 mmHg ( 21 °C)
Product TX1395 is not currently sold in your country. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

3.9 (vs air)

Quality Segment

vapor pressure

41 mmHg ( 21 °C)

assay

≥95% (GC)

form

liquid

autoignition temp.

745 °F

expl. lim.

6 %

color

APHA: ≤10

refractive index

n20/D 1.391 (lit.)

pH

7

bp

98-99 °C (lit.)

mp

−107 °C (lit.)

transition temp

flash point -12 °C

solubility

0.56 mg/L

density

0.692 g/mL at 25 °C (lit.)

shipped in

ambient

storage temp.

room temp

SMILES string

CC(C)CC(C)(C)C

InChI

1S/C8H18/c1-7(2)6-8(3,4)5/h7H,6H2,1-5H3

InChI key

NHTMVDHEPJAVLT-UHFFFAOYSA-N

General description

EMPARTA solvents are designed for routine chemical synthesis, drying, purification, and critical labware cleaning in quality control environments. Engineered for precision, they deliver consistently reproducible results essential for rigorous validation and documentation.

Exceptional Quality Standards: These solvents meet or exceed ACS specifications, providing assurance of high quality that is crucial for accurate testing and analysis in quality control.

Consistent Reproducibility: Designed for precision, the solvents ensure that results can be reliably reproduced, which is vital for maintaining compliance and integrity in quality control processes.

Versatile Applications: Suitable for a variety of QC tasks, including analytical testing, method validation, and testing USP/NF monographs, these solvents support a comprehensive approach to quality assurance in laboratory settings.

Application

Practical

Analysis Note

Assay (GC): 95% min
Color (APHA): 10 max
Form: Clear liquid
Infrared Spectrum: Conforms to standard


Still not finding the right product?

Explore all of our products under 2,2,4-Trimethylpentane


signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

10.4 °F - closed cup

flash_point_c

-12 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters